Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Antibody (CF 6J12), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064440C
Description
Chlamydia Monoclonal antibody specifically detects Chlamydia in Bacteria samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Chlamydia | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Bacteria | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
CF 6J12 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Chlamydia trachomatis | |
Elementary bodies from C. trachomatis strain SAF2. | |
0.5 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction