Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia Ebs Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017580 0.1 mL
NB10064442 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017580 Supplier Novus Biologicals Supplier No. NB100644420.1ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Goat Polyclonal Antibody

Chlamydia Ebs Polyclonal specifically detects Chlamydia Ebs in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Chlamydia Ebs
Applications Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Host Species Goat
Immunogen Chlamydia trachomatis, strain L2, and other serovar groups.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with all antigens of strain L2 and purified elementary bodies of serovars A-K and L1-L3. Cross reactivity has been observed with C. psittacii and C. pneumoniae (TWAR). It does not react with HEp-2 cells or egg yolk sac proteins.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.