Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia LPS MAb Mouse anti-All, Unconjugated, Clone: Clone 2, MilliporeSigma™
Mouse Monoclonal Antibody
$819.52 - $8192.03
Specifications
Antigen | Chlamydia LPS MAb |
---|---|
Clone | Clone 2 |
Applications | ELISA, Immunodiffusion, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABX84941MI
|
MilliporeSigma
MABX84941KC |
1 mg |
Each for $819.52
|
|
|||||
MABX89410MI
|
MilliporeSigma
MABX849410KC |
10 mg |
Each for $8,192.03
|
|
|||||
Description
Chlamydia LPS MAb Monoclonal antibody specifically detects Chlamydia LPS MAb in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Chlamydia LPS MAb | |
ELISA, Immunodiffusion, Western Blot | |
Unconjugated | |
Infectious Diseases | |
Anti-Chlamydia LPS | |
IgG2b | |
Affinity Purified | |
Specific for S. paratyphi |
Clone 2 | |
Monoclonal | |
Mouse | |
All species | |
Chlamydia Trachomatis LPS | |
Primary | |
2 to 8°C. For long-term storage, freeze at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title