Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlamydia LPS Monoclonal Antibody (B410F), FITC
GREENER_CHOICE

Catalog No. MA17339
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17339 Supplier Invitrogen™ Supplier No. MA17339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7339 antibody reacts with Chlamydia trachomatis LPS in bacterial and infected tissue samples and is reactive with all serovars tested. MA1-7339 also cross reacts with C pneumoniae and C.psittaci. MA1-7339 has been successfully used in ELISA, Immunofluorescence and Immunohistochemical applications. The MA1-7339 antibody was raised against group-specific lipopolysaccharide.

Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia LPS
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone B410F
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Chlamydia Lipopolysaccharide
Host Species Mouse
Immunogen Lipopolysaccharide of Chlamydia trachomatis.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.