Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chlamydia LPS Monoclonal Antibody (H85A)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518187
Description
This antibody cross-reacts with LPS from C. pneumoniae & C. psittaci. Purity is >90%. Recommended pairs for ELISA: MA5-18187 (capture) and MA5-18184 (conjugated, detection).
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Specifications
Chlamydia LPS | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2b |
ELISA | |
H85A | |
Unconjugated | |
Chlamydia Lipopolysaccharide | |
Chlamydia sp. native protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction