Learn More
Invitrogen™ Chlamydia pneumoniae Monoclonal Antibody (1696)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547485
Description
Specificity: Specific for C. pneumoniae (TWAR). Not reactive with Chlamydia trachomatis, Klebsiella pneumoniae, Influenza A, Influenza B, Adenovirus, Mycoplasma pneumoniae, Parainfluenza type 1, Parainfluenza type 2, Parainfluenza type 3, Respiratory Syncytial virus, Legionella pneumoniae, Haemophilus influenzae type B, Bordetella pertussis, Streptococcus pneumoniae, and group A Streptococcus. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Chlamydia was classified in a separate order (Chlamydiales) because of its unique developmental cycle. Chlamydia may be found in the form of an elementary body and a reticulate body and Chlamydia species have genomes of approximately 1 to 1.3 megabases in length. Chlamydia infections are the most common bacterial sexually transmitted infections in humans and are the leading cause of infectious blindness worldwide.
Specifications
Chlamydia pneumoniae | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry | |
1696 | |
Unconjugated | |
C. pneumoniae | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.