Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlamydia trachomatis EBs Polyclonal Antibody, Biotin

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA173072

Catalog No. PIPA173072


Only null left
Add to Cart

Description

Description

The PA1-73072 antibody reacts with Chlamydia trachomatis EBs. PA1-73072 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73072 antibody was raised against purified elementary bodies, disrupted (L2 + other serogroups). Recommended storage of less than 6 months at 2-8ºC. For long term storage, aliquot and freeze/.

Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chlamydia trachomatis EBs
Polyclonal
Biotin
C. trachomatis; C. trachomatis EBS; Chlamydia EB; Chlamydia EBs
Purified elementary bodies, disrupted (L2 + other serogroups).
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry, Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG Fraction
RUO
Bacteria
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.