Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/236), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066402AF350
Description
Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Chlamydia Trachomatis Lps | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Native | |
Primary | |
Bacteria | |
IgG1 |
Western Blot, ELISA | |
1312/236 | |
Western Blot, ELISA | |
Mouse | |
0.1 mL | |
In a simple ELISA this is reactive with 15 serovars of C. trachomatis. The epitope has been shown by Western blotting to be present in LPS. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction