Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Trachomatis Lps Antibody (1312/236) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066402
Description
Chlamydia Trachomatis Lps Monoclonal specifically detects Chlamydia Trachomatis Lps in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Chlamydia Trachomatis Lps | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Native | |
0.1 mg | |
Primary | |
Bacteria | |
Purified |
Western Blot, ELISA | |
1312/236 | |
Unconjugated | |
Mouse | |
Protein A purified | |
RUO | |
In a simple ELISA this is reactive with 15 serovars of C. trachomatis. The epitope has been shown by Western blotting to be present in LPS. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction