Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia Trachomatis Momp Antibody (1297/143), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066403FR
Description
Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Chlamydia Trachomatis Momp | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
O175_03780 | |
Native type E MOMP | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Western Blot, ELISA | |
1297/143 | |
Western Blot, ELISA | |
Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2 | |
Mouse | |
Protein A purified | |
RUO | |
Reacts with MOMP from C. trachomatis. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction