Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlamydia trachomatis MOMP Monoclonal Antibody (CL12-666.33.3), Invitrogen™

Catalog No. PIMA546112
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546112 100 μg
01-671-591 500 μg
PIMA546057 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546112 Supplier Invitrogen™ Supplier No. MA546112
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10660 detects Chlamydia trachomatis MOMP from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10660 has been successfully used in ELISA applications. The MA1-10660 immunogen is C. trachomatis elementary bodies.

Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia trachomatis MOMP
Applications ELISA
Classification Monoclonal
Clone CL12-666.33.3
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C. trachomatis MOMP; Chlamydia MOMP
Host Species Mouse
Immunogen C. trachomatis elementary bodies.
Purification Method PEG precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.