Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlamydia trachomatis MOMP Monoclonal Antibody (CL12-741.24.5.5), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | Chlamydia trachomatis MOMP |
---|---|
Clone | CL12-741.24.5.5 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7579
![]() ![]() |
Leinco Technologies Inc
15140100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-7580
![]() ![]() |
Leinco Technologies Inc
15140500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-7578
![]() ![]() |
Leinco Technologies Inc
151401000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
Chlamydia trachomatis MOMP Monoclonal antibody specifically detects Chlamydia trachomatis MOMP in Bacteria samples. It is validated for ELISASpecifications
Chlamydia trachomatis MOMP | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
C. trachomatis elementary bodies, L2 serovar | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
CL12-741.24.5.5 | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
C. trachomatis MOMP; Chlamydia MOMP | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title