Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlamydia trachomatis OmcB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117552
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117552 Supplier Invitrogen™ Supplier No. PA5117552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

In elementary bodies (EBs, the infectious stage, which is able to survive outside the host cell) provides the structural integrity of the outer envelope through disulfide cross-links with the small cysteine-rich protein and the major outer membrane protein. It has been described in publications as the Sarkosyl-insoluble COMC (Chlamydia outer membrane complex), and serves as the functional equivalent of peptidoglycan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia trachomatis OmcB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene omcB
Gene Accession No. P0CC04
Gene Symbols omcB
Host Species Rabbit
Immunogen Recombinant Chlamydia trachomatis Large cysteine-rich periplasmic protein OmcB protein (15-243aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 884223
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.