Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chlamydia trachomatis serovar L2 ompA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117609 Supplier Invitrogen™ Supplier No. PA5117609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

ompA acts as a fimbriae subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydia trachomatis serovar L2 ompA
Applications Western Blot
Classification Polyclonal
Concentration 3.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ompA
Gene Accession No. P0A911
Gene Alias Fimbrin; ompP5; Outer membrane protein P5
Gene Symbols ompA
Host Species Rabbit
Immunogen Recombinant Chlamydia trachomatis serovar L2 Major outer membrane porin protein (23-394aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 917123
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.