Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydial LPS, Mouse anti-Bacteria, Clone: EVI-H1, Millipore Sigma™

Catalog No. MABF2107100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2107100 100 μg
MABF210725 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2107100 Supplier MilliporeSigma Supplier No. MABF2107100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Chlamydial LPS, clone EVI-H1, Cat. No. MABF2107, is a mouse monoclonal antibody that detects Chlamydial lipopolysaccharide (LPS) and has been tested for use in Electron Microscopy, Immunocytochemistry, Immunofluorescence, and Western Blotting.

Chlamydia trachomatis is a Gram-negative bacterium that is responsible to sexually transmitted diseases leading to pelvic inflammatory disease, ectopic pregnancy, infertility, and outbreaks of trachoma-associated blindness and lymphogranuloma venereum (LGV). Chlamydia trachomatis consists of eighteen different serological variants (serovars) that include a few subvariants. These are identified based on serological reactivity of the epitopes on their outer membrane. Intracellularly chlamydia replicates within a vacuole. Chlamydia infection is initiated with the expression of a chlamydial early gene product(s), which isolate the inclusion from the endocytic-lysosomal pathway and makes it fusogenic with sphingomyelin-containing exocytic vesicles. This change in vesicular interaction allows the delivery of the vacuole to the peri-Golgi region of the host cell. Antigens from all members of the Chlamydia genus display heat resistance and sensitivity to oxidation by sodium periodate. Lipopolysaccharides (LPS) of Chlamydia consists of at least three antigen domains, two of which are shared by the LPS of certain other gram-negative organisms and one that is unique only to chlamydial LPS. It has been reported that the quantitative yields of chlamydial LPS can be achieved when elementary bodies (EBs) are first reduced and alkylated prior to extraction with hot phenol-water. (Ref.: Caldwell, HD., and Hitchcock, PJ. (1984). Infect. Immun. (44(2); 306-314; Scidmore, MA., et al. (1996). Infect. Immun. 64(12); 5366-5372; Turingan, RS et al. (2017). PLoS ONE 12(5): e0178653).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydial LPS
Applications Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot
Classification Monoclonal
Clone EVI-H1
Conjugate Unconjugated
Dilution Immunofluorescence Analysis: A representative lot detected Chlamydial LPS in Immunofluorescence applications (Su, H., et. al. (2000). Infect Immun. 68(1):192-6; Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).Immunocytochemistry Analysis: A representative lot detected Chlamydial LPS in Immunocytochemistry applications (Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).Electron Microscopy Analysis: A representative lot detected Chlamydial LPS with Trasnsmission Electron Microscopy applications (Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Chlamydial lipopolysaccharide;Chlamydia trachomatis LPS
Host Species Mouse
Immunogen Formalin-killed elementary bodies of the Chlamydia trachomatis L2 serovar.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone EVH-H1 detects lipopolysaccharide from Chlamydia genus.
Target Species Bacteria
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.