Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydial MOMP, Mouse anti-Bacteria, Clone: L21-10, Millipore Sigma™

Catalog No. MABF216625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF216625 25 μg
MABF2166100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF216625 Supplier MilliporeSigma Supplier No. MABF216625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Chlamydial MOMP, clone L21-10, Cat. No. MABF2166, is a mouse monoclonal antibody that detects Chlamydial major outer membrane protein (MOMP) and has been tested for use in Dot Blot, Immunofluorescence, Immunoprecipitation, and Western Blotting.

Chlamydia trachomatis is a Gram-negative bacterium that is responsible for sexually transmitted diseases leading to pelvic inflammatory disease, ectopic pregnancy, infertility, and outbreaks of trachoma-associated blindness and lymphogranuloma venereum (LGV). Chlamydia trachomatis consists of eighteen different serological variants (serovars) that include a few subvariants. These are identified based on serological reactivity of the epitopes on their outer membrane. Intracellularly chlamydia replicates within a vacuole. Chlamydia infection is initiated with the expression of a chlamydial early gene product(s), which isolate the inclusion from the endocytic-lysosomal pathway and makes it fusogenic with sphingomyelin-containing exocytic vesicles. This change in vesicular interaction allows the delivery of the vacuole to the peri-Golgi region of the host cell. Antigens from all members of the Chlamydia genus display heat resistance and sensitivity to oxidation by sodium periodate. Clone L21-10 specifically detects major outer membrane protein (MOMP) from Chlamydia genus. It displays strong reactivity with Chlamydial serovars D and L3 and weak reactivity with Ba, E, G, L1, and L2. MOMP, a cysteine-rich molecule, plays a vital role in maintaining the structural and functional properties of the chlamydial outer membrane. It contains determinants that confer type, subspecies, and species antigenic properties to the protein. (Ref.: Zhang, YX et al., (1987). J. Immunol. 138(2); 575-581).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chlamydial MOMP
Applications Dot Blot, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone L21-10
Conjugate Unconjugated
Dilution Immunoprecipitation Analysis: A representative lot immunoprecipiated Chlamydial MOMP in Immunoprecipitation applications (Zhang, Y.X., et. al. (1987). J Immunol. 138(2):575-81).Immunofluorescence Analysis: A representative lot detected Chlamydial MOMP in Immunofluorescence applications (Weber, M.M., et. al. (2017). Cell Rep. 19(7):1406-1417; Zhang, Y.X., et. al. (1987). J Immunol. 138(2):575-81).Western Blotting Analysis: A representative lot detected Chlamydial MOMP in Western Blotting applications (Baehr, W., et. al. (1988). Proc Natl Acad Sci USA. 85(11):4000-4; Zhang, Y.X., et. al. (1987). J Immunol. 138(2):575-81).Dot Blot Analysis: A representative lot detected Chlamydial MOMP in Dot Blot applications (Zhang, Y.X., et. al. (1987). J Immunol. 138(2):575-81).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Chlamydia major membrane protein
Gene Symbols N/A
Host Species Mouse
Immunogen Formalin killed Chlamydia trachomatis (L2/LGV-434).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone L21-10 specifically detects outer membrane protein from Chlamydia genus. It displays strong reactivity with serovars D and L3 and weak reactivity with Ba, E, G, L1, and L2.
Target Species Bacteria
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.