Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHLI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144520 Supplier Invitrogen™ Supplier No. PA5144520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHLI1 Polyclonal Antibody for Western Blot, ELISA

Involved in chlorophyll biosynthesis. Catalyzes the insertion of magnesium ion into protoporphyrin IX to yield Mg-protoporphyrin IX. The magnesium-chelatase is a complex of three subunits, CHLI, CHLD and CHLH. The reaction takes place in two steps, with an ATP-dependent activation followed by an ATP-dependent chelation step. Possesses high affinity for ATP and may play a major role in chlorophyll biosynthesis. Does not bind abscisic acid (ABA), but is a positive regulator of ABA signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHLI1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.403 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CHLI1
Gene Accession No. P16127
Gene Alias CH42; CS
Gene Symbols CHLI1
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana CHLI1 protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 827580
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.