Learn More
Chloramine-T trihydrate, ACS, 98.0-103.0%
Description
Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 167°C to 170°C |
| Physical Form | Crystalline |
| Quantity | 100 g |
| UN Number | UN3263 |
| Beilstein | 3924168 |
| Flash Point | 192°C (377°F) |
| Merck Index | 14,2075 |
| Solubility Information | Soluble in water and ethyl alcohol. Insoluble in benzene and ethers. |
| Formula Weight | 281.69 (227.67 Anhydrous) |
| Percent Purity | 98.0 to 103.0% |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H334-H335 |
| Hazard Statement | GHS H Statement H334-H314-H318-H302 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S. |
| EINECSNumber | 204-854-7 |
| RTECSNumber | XT5616800 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.