Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol Acetyltransferase Antibody (CAT-1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20129A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20129A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20129A32 Supplier Novus Biologicals Supplier No. NBP250129AF532

Mouse Monoclonal Antibody

Chloramphenicol Acetyltransferase Monoclonal specifically detects Chloramphenicol Acetyltransferase in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Chloramphenicol Acetyltransferase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CAT-1
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CAT, CAT II, CATB, CatD, Chloramphenicol acetyltransferase 2, Chloramphenicol acetyltransferase II, cmlA
Host Species Mouse
Immunogen Full length Chloramphenicol Acetyltransferase
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.