Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloramphenicol Acetyltransferase Antibody (CAT-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Chloramphenicol Acetyltransferase |
---|---|
Clone | CAT-1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Description
Chloramphenicol Acetyltransferase Monoclonal antibody specifically detects Chloramphenicol Acetyltransferase in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Chloramphenicol Acetyltransferase | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Epitope Tags | |
PBS | |
Full length Chloramphenicol Acetyltransferase | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CAT-1 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Non-species specific | |
CAT, CAT II, CATB, CatD, Chloramphenicol acetyltransferase 2, Chloramphenicol acetyltransferase II, cmlA | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title