Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloramphenicol Antibody - BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB100624570.1ML
Description
Chloramphenicol Polyclonal specifically detects Chloramphenicol in Non-species specific samples. It is validated for ELISA.Specifications
Chloramphenicol | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Chloramphenicol base conjugated via its amino group to KLH using phenylenediisothiocyanate | |
0.1 mL | |
Primary | |
Non-species specific | |
Purified |
ELISA | |
5.0 mg/mL | |
ELISA 1:100-1:2000 | |
Sheep | |
IgG purified | |
RUO | |
This product recognises chloramphenicol, a broad-spectrum antimicrobial that inhibits protein synthesis. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction