Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Dihydrochloride 96.0+%, TCI America™
SDP

Catalog No. C125425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C125425G 25 g
C12545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C125425G Supplier TCI America Supplier No. C125425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3697-42-5
Molecular Formula C22H32Cl4N10
Molecular Weight (g/mol) 578.368
MDL Number MFCD00068998
InChI Key WJLVQTJZDCGNJN-UHFFFAOYSA-N
Synonym 1,1′C-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Dihydrochloride
PubChem CID 9571016
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;dihydrochloride
SMILES C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.Cl.Cl

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 578.37
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorhexidine Dihydrochloride
RTECSNumber XA5285000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.