Alfa Aesar™ Chloride, Ion chromatography standard solution, Specpure™, Cl- 1000µg/mL

Manufacturer: Alfa Aesar™ 35551AE

Quantity 100mL

 View more versions of this product

Catalog No. AA35551AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00143949
    Chemical Name or Material Chloride Ion chromatography standard solution, Cl- 1000μg/mL
    Grade Specpure™

    Specifications

    Physical Form Liquid
    MDL Number MFCD00143949
    Quantity 100mL
    TSCA TSCA
    Chemical Name or Material Chloride Ion chromatography standard solution, Cl- 1000μg/mL
    Grade Specpure™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit