Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Standard, 0.100 N (N/10), 100 mEq/L NaCl, Ricca Chemical
SDP

Catalog No. 19601
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7647-14-5, 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.100Normal
CAS Min % 97.0, 0.57
CAS Max % 100.0, 0.59
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 4 L
Chemical Name or Material Chloride Standard, 100mEq/L NaCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.