Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Standard, 10 ppm Cl-, Ricca Chemical
SDP

Catalog No. 1942116 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L

Specifications

Chemical Name or Material CHLORIDE STANDARD
CAS 12125-02-9
Molecular Formula ClH4N
MDL Number MFCD00011420
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name amine hydrochloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Grade Laboratory
Packaging Natural Poly Bottle
pH 6.7 to 7.3
Quantity 500 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.