Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloride Standard, 500 ppm Cl- (0.0141 N), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 98.0, 0.08 |
CAS Max % | 100.0, 0.08 |
Equivalence | 1 mL = 0.5 mg Cl |
Physical Form | Liquid |
Color | Colorless |
Boiling Point | 100°C |
Quantity | 1 L |
Chemical Name or Material | Chloride Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction