Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorine Water, Saturated, Approximately 0.3% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Safety-coated Glass Bottle
Specifications
Specifications
| Color | Green-Yellow |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Grade | Laboratory |
| Packaging | Safety-coated Glass Bottle |
| Solvent or Matrix | Saturated Aqueous Solution |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | approximately 0.3% (w/v) |
| Chemical Name or Material | Chlorine Water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction