Learn More
Eisco™ Chlorite Mineral Specimen
SCI_ED
Catalog No. S26674 Shop All Eisco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S26674 10/Pk.
1 options
Catalog No. S26674 Supplier Eisco™ Supplier No. ESNG0068PK10
Add to Cart
Edge
Add to Cart
Chlorite is a metamorphic mineral that has undergone light alteration.
  • Greener color and near-soapy feel of chlorite distinguishes it from other phyllosilicates
  • presence of water distinguishes it chemically from other phyllosillcates
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Forms when hot water alters the chemistry of an existing mineral
  • During metasomatism, phyllosilicate minerals may become hydrated and form chlorite
  • This sample is ideal for lessons on metamorphism, mineral alteration, and general identification exercises
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Chlorite