Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chloro(1,5-hexadiene)rhodium(I) dimer, Thermo Scientific™

Catalog No. AC377920010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 32965-49-4
Molecular Formula C12H20Cl2Rh2
Molecular Weight (g/mol) 441.02
MDL Number MFCD00064595

Specifications

Melting Point 118.0°C to 120.0°C
Color Golden
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Formula Weight 441.02
Physical Form Crystalline Powder
Chemical Name or Material chloro(1, 5-hexadiene)rhodium(I) dimer
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.