Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(chloromethyl)dimethylsilane, 98%, AcroSeal™

Catalog No. ac434021000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-57-9
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.08
MDL Number MFCD00000875
InChI Key ITKVLPYNJQOCPW-UHFFFAOYSA-N
Synonym chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
PubChem CID 74393
IUPAC Name chloro-(chloromethyl)-dimethylsilane
SMILES C[Si](C)(Cl)CCl

Specifications

Color Colorless to Straw
Density 1.0800g/mL
Boiling Point 114.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4360 to 1.4380
Linear Formula ClCH2Si(CH3)2Cl
Quantity 100 mL
Specific Gravity 1.08
Solubility Information Solubility in water: reacts
Formula Weight 143.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloro(chloromethyl)dimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 217-006-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.