Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroacetic acid, 99%
Click to view available options
Quantity:
250 g
1000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.
Solubility
Soluble in water, methanol, acetone, diethyl ether, benzene, chloroform and ethanol.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.
Solubility
Soluble in water, methanol, acetone, diethyl ether, benzene, chloroform and ethanol.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | 60°C to 63°C |
| Density | 1.404 |
| pH | <1.0 |
| Boiling Point | 189°C |
| Flash Point | 126°C (258°F) |
| Odor | Pungent |
| Linear Formula | ClCH2CO2H |
| Quantity | 250 g |
| UN Number | UN1751 |
| Beilstein | 605438 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311-H314-H330-H335 |
| Hazard Statement | GHS H Statement H301-H311-H331-H314 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETIC ACID, SOLID |
| EINECSNumber | 201-178-4 |
| RTECSNumber | AF8575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction