Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetic acid, 99+%, ACS reagent, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
79-11-8
C2H3ClO2
94.5
FOCAUTSVDIKZOP-UHFFFAOYSA-N
chloroacetic acid,monochloroacetic acid,chloracetic acid,acetic acid, chloro,acide chloracetique,chloroethanoic acid,monochloroethanoic acid,monochloracetic acid,monochloorazijnzuur,monochloressigsaeure
300
CHEBI:27869
2-chloroacetic acid
C(C(=O)O)Cl
79-11-8
C2H3ClO2
94.5
FOCAUTSVDIKZOP-UHFFFAOYSA-N
chloroacetic acid,monochloroacetic acid,chloracetic acid,acetic acid, chloro,acide chloracetique,chloroethanoic acid,monochloroethanoic acid,monochloracetic acid,monochloorazijnzuur,monochloressigsaeure
300
CHEBI:27869
2-chloroacetic acid
C(C(=O)O)Cl
Chloride 0.01% max
Sulfate 0.02% max
Insoluble Matter 0.01% max
Viscosity 1.29 Pas (100°C)
Specific Gravity 1.58
Packaging Plastic bottle
Quantity 100g
Flash Point 126°C
Boiling Point 189.0°C
Density 1.5800g/mL
Melting Point 61.0°C to 63.0°C
Assay Percent Range 99+%
Linear Formula ClCH2CO2H
Merck Index 15,2112
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
Formula Weight 94.5
Percent Purity 99+%
Grade ACS Reagent
Chemical Name or Material Chloroacetic acid
Show More Show Less

GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins

GHS Signal Word: Danger

EINECSNumber : 201-178-4