Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetic acid, 99%, flakes

Catalog No. AC108510025
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
4 kg
Packaging:
Plastic bottle

CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.5
MDL Number MFCD00002683
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Decomposition Information 250°C
Melting Point 61.0°C to 63.0°C
Color White
Density 1.5800g/mL
Boiling Point 189.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClCH2CO2H
Quantity 2.5 kg
Beilstein 02, 194
Specific Gravity 1.58
Merck Index 15, 2112
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
Viscosity 1.29 Pas (100°C)
Vapor Pressure 0.75 mmHg at 20°C
Formula Weight 94.5
Percent Purity 99%
Physical Form Crystalline Flakes
Chemical Name or Material Chloroacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins
EINECSNumber 201-178-4
RTECSNumber AF8575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.