Learn More
Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil
Description
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
Solubility
Soluble in water (124g/L), ether and chloroform.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.

Specifications
Specifications
Melting Point | -45°C |
Density | 1.161 |
pH | 4.3 |
Boiling Point | 118°C to 120°C |
Flash Point | 40°C (104°F) |
Odor | Pungent |
Assay Percent Range | 95%, Stabilized with 0.1% Epoxidized Soybean Oil |
Refractive Index | 1.434 |
Linear Formula | ClCH2COCH3 |
Quantity | 1000 g |
Show More |
Safety and Handling
Hazard Category | H226-H301+H311-H315-H319-H330-H335 |
Hazard Statement | GHS H Statement H301-H310-H330-H314-H318-H226 Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETONE, STABILIZED |
EINECSNumber | 201-161-1 |
RTECSNumber | UC0700000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.