Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetonitrile, 98+%

Catalog No. p-85888
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle

CAS: 107-14-2 | C2H2ClN | 75.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-14-2
Molecular Formula C2H2ClN
Molecular Weight (g/mol) 75.5
MDL Number MFCD00001885
InChI Key RENMDAKOXSCIGH-UHFFFAOYSA-N
Synonym chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
PubChem CID 7856
ChEBI CHEBI:82416
IUPAC Name 2-chloroacetonitrile
SMILES C(C#N)Cl

Specifications

Color Colorless
Density 1.1890g/mL
Boiling Point 124.0°C to 126.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4240
Linear Formula ClCH2CN
Quantity 100 mL
Beilstein 02,201
Fieser 01,129; 06,102
Specific Gravity 1.189
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, diethyl ether, hydrocarbons
Formula Weight 75.5
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Chloroacetonitrile, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 203-467-0
RTECSNumber AL8225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.