Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, 99.5%, Acros Organics™

Click to view available options
Quantity:
100mL
1L
108-90-7
C6H5Cl
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl
108-90-7
C6H5Cl
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl
Linear Formula C6H5Cl
Solubility Information Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 112.56
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Density 1.1075g/mL
Refractive Index 1.5230 to 1.5250
Solubility Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Specific Gravity 1.1075
Water 0.005% max.
Flash Point 23°C
Viscosity 0.8 mPa.s (20°C)
Boiling Point 131.0°C
Melting Point -45.0°C
Quantity 100mL
Chemical Name or Material Chlorobenzene
Show More Show Less

GHS H Statement Toxic to aquatic life with long lasting effects. Harmful if inhaled. Flammable liquid and vapor. Causes skin irritation. May cause drowsiness or dizziness.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling.

Warning