Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, ≥99.5%, ACS reagent

Catalog No. ac404490010
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Density 1.1075g/mL
Quantity 1 kg
Infrared Spectrum Authentic
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.5230 to 1.5250 (20°C, 589nm)
Assay Percent Range 99.6%
Residue after Evaporation 0.02% max.
Titratable Acid 0.004 meq/g max.
Linear Formula C6H5Cl
Vapor Pressure Vapor Pressure: 12mbar at 20°C
UN Number 1134
Beilstein 05, 199
Merck Index 15, 2122
Formula Weight 112.56
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material Chlorobenzene, For analysis ACS
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.