Learn More
Chlorobenzene, ACS, 99.5%, Thermo Scientific Chemicals
Description
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -45°C |
Color | Colorless |
Density | 1.106 g/mL |
Boiling Point | 132°C |
Quantity | 500 mL |
Flash Point | 28°C (82°F) |
Odor | Aromatic |
Physical Form | Liquid |
Refractive Index | 1.524 |
UN Number | UN1134 |
Show More |
Safety and Handling
Hazard Category | H226-H315-H332 |
Hazard Statement | GHS H Statement H226-H332 Flammable liquid and vapor. Harmful if inhaled. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE |
EINECSNumber | 203-628-5 |
RTECSNumber | CZ0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.