Learn More
Chlorobenzene, HPLC Grade, 99.5%
Description
Chlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -45°C |
Color | Colorless |
Density | 1.106 g/mL |
Boiling Point | 132°C |
Quantity | 4 x 1 L |
Flash Point | 28°C (82°F) |
Odor | Aromatic |
Physical Form | Liquid |
Refractive Index | 1.524 |
UN Number | UN1134 |
Show More |
Safety and Handling
Hazard Category | H226-H315-H332 |
Hazard Statement | GHS H Statement H226-H332 Flammable liquid and vapor. Harmful if inhaled. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE |
EINECSNumber | 203-628-5 |
RTECSNumber | CZ0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.