Chlorobenzene, Spectrophotometric Grade, 99.9%, Alfa Aesar™

Manufacturer: Alfa Aesar 22921M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA22921M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-90-7
    Physical Form Liquid
    Assay Percent Range 99.9%
    Molecular Formula C6H5Cl
    Formula Weight 112.56
    MDL Number MFCD00000530
    Chemical Name or Material Chlorobenzene
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 112.56
    Quantity 4 x 1L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: III
    EINECS Number 203-628-5
    Physical Form Liquid
    Odor Aromatic
    Vapor Pressure 12 hPa (9mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range 99.9%
    Molecular Formula C6H5Cl
    MDL Number MFCD00000530
    CAS 108-90-7
    Health Hazard 1 H226-H332-H411: Flammable liquid and vapour. Harmful if inhaled. Toxic to aquatic life with long lasting effects.
    Chemical Name or Material Chlorobenzene
    Grade Spectrophotometric grade

    H226-H332-H411: Flammable liquid and vapour. Harmful if inhaled. Toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit