Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobutanol, Anhydrous, NF, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601602 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

CH123, 57-15-8, C4H7Cl3O

Spectrum™ Chemical Chlorobutanol, Anhydrous, NF is a very effective preservative in many pharmaceutical products for eyes, ears and nose, dental preparations. The anhydrous form is useful in formulations where the absence of water is desired. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-15-8
Molecular Formula C4H7Cl3O
Molecular Weight (g/mol) 177.45
InChI Key OSASVXMJTNOKOY-UHFFFAOYSA-N
IUPAC Name 1,1,1-trichloro-2-methylpropan-2-ol
SMILES CC(C)(O)C(Cl)(Cl)Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98-100.5%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.