Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorocyclopentane, 99%

Catalog No. p-7027947
Click to view available options
Quantity:
100 g
500 g

CAS: 930-28-9 | C5H9Cl | 104.577 g/mol

Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. 760 mg/L.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-28-9
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00001360
InChI Key NDTCXABJQNJPCF-UHFFFAOYSA-N
Synonym cyclopentane, chloro, cyclopentyl chloride, chloroclopentane, cyclopentylchloride, unii-to3f41cl9a, 1-chlorocyclopentane, to3f41cl9a, cyclopentylchlorid, chloro-cyclopentan, chlorocyclopenetane
PubChem CID 70252
IUPAC Name chlorocyclopentane
SMILES C1CCC(C1)Cl

Specifications

Melting Point -94°C
Density 1.003
Boiling Point 113°C to 114°C
Flash Point 15°C (59°F)
Refractive Index 1.4512
Quantity 100 g
UN Number UN1993
Beilstein 1900557
Solubility Information Soluble in water. 760mg/L.
Formula Weight 104.58
Percent Purity 99%
Chemical Name or Material Chlorocyclopentane
Show More Show Less
Hazard Category H225-H315-H319
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-212-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.