Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethyl(3-phenylpropyl)silane 97.0+%, TCI America™
SDP

Catalog No. C148825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C148825ML 25 mL
C14885ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C148825ML Supplier TCI America Supplier No. C148825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17146-09-7
Molecular Formula C11H17ClSi
Molecular Weight (g/mol) 212.79
MDL Number MFCD00049281
InChI Key ASSMBLOISZSMMP-UHFFFAOYSA-N
PubChem CID 86971
IUPAC Name chlorodimethyl(3-phenylpropyl)silane
SMILES C[Si](C)(Cl)CCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 121°C
Quantity 25 mL
UN Number 2987
Formula Weight 212.79
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethyl(3-phenylpropyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.