Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethylpropylsilane 99.0+%, TCI America™
SDP

Catalog No. D159025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D159025ML 25 mL
D15905ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D159025ML Supplier TCI America Supplier No. D159025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17477-29-1
Molecular Formula C5H13ClSi
Molecular Weight (g/mol) 136.694
MDL Number MFCD00039301
InChI Key HXVPUKPVLPTVCQ-UHFFFAOYSA-N
Synonym propyldimethylchlorosilane, chlorodimethylpropylsilane, n-propyldimethylchlorosilane, silane, chlorodimethylpropyl, dimethylpropylchlorosilane, chlorodimethyl propyl silane, chlorodimethyl-n-propylsilane
PubChem CID 87130
IUPAC Name chloro-dimethyl-propylsilane
SMILES CCC[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 116°C
Quantity 25 mL
UN Number 2986
Formula Weight 136.69
Percent Purity ≥99.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material Chlorodimethylpropylsilane [Dimethylpropylsilylating Agent]
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.