Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(Dimethylsulfide)Gold(I)

Catalog No. 4207271G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-072-71G 1 g
42-072-7250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 42-072-71G Supplier Sigma Aldrich Supplier No. 4207271G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloro(dimethylsulfide)gold(I) can be used to synthesize a gold(I)-catalyst that can effectively catalyze intermolecular [2+2] cycloaddition of terminal alkynes with substituted alkenes to form substituted cyclobutenes under mild reaction conditions.It can also be used to synthesize:chloro(trimesitylphosphine)gold(I)gold(I) N-heterocyclic carbene (NHC) complexesbis(carbene)gold(I) complex of ferrocenyl complexesgold(I) acetylide complexes

Specifications

CAS 29892-37-3
Molecular Formula C2H6AuClS
Linear Formula (CH3)2SAuCl
MDL Number MFCD00192090
Quantity 1 g
Synonym (Dimethylsulfide)gold(I) chloride
Molecular Weight (g/mol) 294.55 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.