Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ≥99.5%, Solstice

Catalog No. 60047876 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
4 x 25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-876 1 L Glass bottle
60-047-878 25 mL Glass bottle
60-047-880 500 mL Glass bottle
60-047-877 2.5 L Glass bottle
60-047-879 4 x 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-047-876 Supplier Solstice Supplier No. C24321L
Add to Cart
Add to Cart

Contains 100-200 ppm amylenes as stabilizer,≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -61.3°C
Color Colorless
Density 1.492g/mL at 25°C
Boiling Point 61°C (1013 hPa)
Odor Sweet
Packaging Glass bottle
Quantity 1 L
UN Number UN1888
Beilstein 1731042
IUPAC Name chloroform
Stabilizer Amylenes
Vapor Density 4.1 (vs air)
Vapor Pressure 160mmHg (20°C)
Formula Weight 119.38g/mol
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Chloroform
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338-P311
DOTInformation 6.1/PGIII
RTECSNumber 200-663-8
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.