Learn More
Chloroform, 99+%, extra pure, stabilized with amylene, ACROS Organics™
Specifications
Assay Percent Range | 99+% |
Linear Formula | CHCl3 |
Merck Index | 15, 2142 |
Solubility Information | Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone. |
Formula Weight | 119.38 |
Physical Form | Liquid |
Percent Purity | ≥99% |
Grade | Extra Pure |
Density | 1.4800g/mL |
Packaging | Glass bottle |
Show More |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning