Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002579
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Histological Use

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Quantity 1 L
Packaging Aluminum Bottle
Specific Gravity 1L = 1.48kg
Formula Weight 119.38
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Chloroform
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.