Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Environmental Grade, 99.8+%, stab. with ethanol

Catalog No. AA40974K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40974K2 1 L
AA40974M1 4 x 1 L
AA40974K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40974K2 Supplier Thermo Scientific Chemicals Supplier No. 040974K2
Add to Cart

Suitable For Pesticide Analysis, Environmental and GC Analysis | CAS: 67-66-3 | CHCl3 | 119.37 g/mol

It is suitable for pesticide analysis, environmental and GC analysis. It is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform, plays an essential role in the RNA isolation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point °C to 63°C
Color Colorless
Appearance Colorless liquid
Density 1.492
Quantity 1 L
Ignition Point 982°C (1800°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.446
Assay Percent Range >99.8%
Vapor Pressure 210 hPa (158mm Hg) at 20°C
Beilstein 1731042
Boiling Point 60.5°C to 61.5°C
Solubility Information Miscible in diethyl ether,oils,ligroin,alcohol,CCl4 and CS2.
IUPAC Name chloroform
Formula Weight 119.38
Grade Environmental
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category H302-H315-H319-H331-H335-H336-H351-H361d-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 200-663-8
RTECSNumber FS9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.