Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform, For gas chromatography ECD and FID SupraSolv™ MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | -63°C |
Color | Colorless |
Density | 1.4g/cm3 at 20°C |
Boiling Point | 61°C |
Quantity | 1 L |
Odor | Sweet |
Packaging | Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 211 hPa (20°C) |
UN Number | 1888 |
Show More |
Safety and Handling
Hazard Category | H302: Harmful if swallowed. |
Hazard Statement | H315: Causes skin irritation. |
Precautionary Statement | H319: Causes serious eye irritation. |
Recommended Storage | 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction